Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human alpha-Synuclein Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB13383100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB13383100 100 μg
MAB13383SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB13383100 Supplier R&D Systems Supplier No. MAB13383100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Synuclein Monoclonal specifically detects alpha-Synuclein in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen alpha-Synuclein
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 970114
Conjugate Unconjugated
Dilution Western Blot 5 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P37840
Gene Alias alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein
Host Species Mouse
Immunogen E. coli-derived recombinant human alpha-Synuclein Met1-Ala140 Accession # P37840
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6622
Test Specificity Detects human alpha-Synuclein in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.