Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human alpha-Synuclein Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB11376100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB11376100 100 μg
MAB11376SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB11376100 Supplier R&D Systems Supplier No. MAB11376100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

alpha-Synuclein Monoclonal antibody specifically detects alpha-Synuclein in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry

Specifications

Antigen alpha-Synuclein
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 1060506
Conjugate Unconjugated
Dilution Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. P37840
Gene Alias alpha-synuclein, I+/--synuclein, MGC110988, NACP, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, PD1, SNCA, synuclein alpha-140, synuclein, alpha (non A4 component of amyloid precursor), Synuclein-alpha, truncated alpha synuclein
Host Species Mouse
Immunogen E. coli -derived human alpha-Synuclein, Met1-Ala140, Accession # P37840
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6622
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.