Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human AMICA DuoSet ELISA
SDP

Catalog No. DY3449 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
15 x 96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY3449 15 x 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY3449 Supplier R&D Systems™ Supplier No. DY3449
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Human AMICA DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human AMICA/JAML.

Specifications

Quantity 15 x 96-well Plates
Conjugate Biotin
Assay Range 234.00 to 15000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 120425
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.