Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AMICA/JAML Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FA34491T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FA34491T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FA34491T100 Supplier R&D Systems Supplier No. FAB34491T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

AMICA/JAML Monoclonal specifically detects AMICA/JAML in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen AMICA/JAML
Applications Flow Cytometry
Classification Monoclonal
Clone 401901
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q86YT9
Gene Alias adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML
Gene Symbols AMICA1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML Leu20-Leu275 Accession # Q86YT9
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120425
Test Specificity Detects human AMICA/JAML in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse AMICA is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.