Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AMICA/JAML Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB34491
This item is not returnable.
View return policy
Description
AMICA/JAML Monoclonal specifically detects AMICA/JAML in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
AMICA/JAML | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Q86YT9 | |
AMICA1 | |
Mouse myeloma cell line NS0-derived recombinant human AMICA/JAML Leu20-Leu275 Accession # Q86YT9 | |
100 μg | |
Primary | |
Detects human AMICA/JAML in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse AMICA is observed. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
401901 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
120425 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction