Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human AMICA/JAML Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF3449
This item is not returnable.
View return policy
Description
AMICA/JAML Polyclonal specifically detects AMICA/JAML in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
AMICA/JAML | |
Polyclonal | |
Biotin | |
Q86YT9 | |
AMICA1 | |
Mouse myeloma cell line NS0-derived recombinant human AMICA Lys20-Lys275 Accession # Q86YT9 | |
50 μg | |
Primary | |
Detects human AMICA in ELISAs and Western blots. In sandwich immunoassays, less than 0.3% cross-reactivity with recombinant mouse AMICA, recombinant human (rh) JAM-A, rhJAM-B, and rhJAM-C is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
adhesion molecule, interacts with CXADR antigen 1, AMICA, AMICA1, CREA7-1, Gm638, JAML | |
Goat | |
Affinity Purified | |
RUO | |
120425 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction