Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AMIGO Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2565
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2565 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2565 Supplier R&D Systems Supplier No. BAF2565
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

AMIGO Polyclonal specifically detects AMIGO in Human samples. It is validated for Western Blot.

Specifications

Antigen AMIGO
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q86WK6
Gene Alias adhesion molecule with Ig-like domain 1, ALI2, Alivin-2, AMIGO1, AMIGO-1, AMIGOamphoterin-induced protein 1, amphoterin-induced gene and ORF, KIAA1163
Gene Symbols AMIGO1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human AMIGO Met1-Gly365 Accession # Q86WK6
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57463
Test Specificity Detects human AMIGO in Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human AMIGO2 is observed and less than 1% cross-reactivity with recombinant mouse AMIGO3 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.