Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aminopeptidase LRAP/ERAP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB3830
This item is not returnable.
View return policy
Description
Aminopeptidase LRAP/ERAP2 Monoclonal specifically detects Aminopeptidase LRAP/ERAP2 in Human samples. It is validated for Western Blot.Specifications
Aminopeptidase LRAP/ERAP2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
Q6P179 | |
ERAP2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase LRAP/ERAP2 Accession # Q6P179 | |
100 μg | |
Primary | |
Detects human Aminopeptidase LRAP/ERAP2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human PILS is observed. | |
Human | |
Purified |
Western Blot | |
3F5 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 3.4.11, EC 3.4.11.3, endoplasmic reticulum aminopeptidase 2, ERAP2, FLJ23633, FLJ23701, FLJ23807, Leukocyte-derived arginine aminopeptidase, LRAPL-RAPEC 3.4.11.- | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
64167 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction