Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Aminopeptidase N/CD13 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3815SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3815SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3815SP Supplier R&D Systems Supplier No. AF3815SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody has been used in 2 publications

Aminopeptidase N/CD13 Polyclonal specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF-ready.

Specifications

Antigen Aminopeptidase N/CD13
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, CyTOF
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 μg/mL, Simple Western 10 μg/mL, Flow Cytometry 0.25 μg/10^6 cells, Immunohistochemistry 0.3-15 μg/mL, Immunoprecipitation 25 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P15144
Gene Alias alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN
Gene Symbols ANPEP
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase N/CD13 Lys69-Lys967 Accession # P15144
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290
Test Specificity Detects human Aminopeptidase N/CD13 in the direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 20% cross-reactivity with recombinant mouse APN is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.