Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aminopeptidase N/CD13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB38152100
This item is not returnable.
View return policy
Description
Aminopeptidase N/CD13 Monoclonal antibody specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
Aminopeptidase N/CD13 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN | |
Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase N/CD13, Lys69-Lys967, Accession # AAA51719 | |
100 μg | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2a |
Flow Cytometry, Immunohistochemistry, CyTOF | |
986002 | |
Flow Cytometry 0.25 ug/106 cells, Immunohistochemistry 0.5-25 ug/mL, CyTOF-ready | |
AAA51719 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
290 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction