Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aminopeptidase O/ONPEP Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4647SP
This item is not returnable.
View return policy
Description
Aminopeptidase O/ONPEP Monoclonal specifically detects Aminopeptidase O/ONPEP in Human samples. It is validated for Immunohistochemistry.Specifications
Aminopeptidase O/ONPEP | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q8N6M6 | |
C9ORF3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Aminopeptidase O/ONPEP Met1-Phe819 Accession # Q8N6M6 | |
25 μg | |
Primary | |
Detects human Aminopeptidase O/ONPEP in direct ELISAs. | |
Human | |
Purified |
Immunohistochemistry | |
562912 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
aminopeptidase O, AOPEP, APO, AP-OFLJ55832, C90RF3, C9orf3, chromosome 9 open reading frame 3, EC 3.4.11.-, FLJ14675, FLJ40923, ONPEP | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84909 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction