Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Aminopeptidase P2/XPNPEP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2490SP
This item is not returnable.
View return policy
Description
Aminopeptidase P2/XPNPEP2 Monoclonal specifically detects Aminopeptidase P2/XPNPEP2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Aminopeptidase P2/XPNPEP2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Aminoacylproline aminopeptidase, APP2, EC 3.4.11.9, mAmP, Membrane-bound aminopeptidase P, Membrane-bound AmP, Membrane-bound APP, xaa-Pro aminopeptidase 2, XPNPEP2, X-Pro aminopeptidase 2, X-prolyl aminopeptidase (aminopeptidase P) 2, membrane-bound | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7512 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Western Blot, Immunoprecipitation | |
391118 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
AAB96394 | |
XPNPEP2 | |
Mouse myeloma cell line NS0-derived recombinant human Aminopeptidase P2/XPNPEP2 His22-Ala650 Accession # AAB96394 | |
25 μg | |
Primary | |
Detects human Aminopeptidase P2/XPNPEP2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse Aminopeptidase P2/XPNPEP2 and no cross-reactivity with recombinant human XPNPEP1 is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction