Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Amyloid beta (aa1-42) Quantikine ELISA Kit

Catalog No. dab142
Click to view available options
Quantity:
1 Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Human Amyloid beta (aa1-42) Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Amyloid beta (aa1-42) Quantikine ELISA Kit quantifies human beta Amyloid in cell culture supernates (100 ul), tissue lysates (10 ul), cerebrospinal fluid (10 ul).

Specifications

Assay Range 7.81 - 500pg/mL
Assay Sensitivity 4.73pg/mL
Conjugate HRP
Cross Reactivity Cross-reactivity observed with 1 or more available related molecules. <50% cross-species reactivity observed with species tested.
Sample Type Cell Culture Supernatant, Cerebrospinal Fluid, Tissue Lysates
Specificity Natural Human Amyloid beta (aa1-42)
For Use With (Application) ELISA
Gene ID (Entrez) 351 (Human); 11820 (Mouse)
Quantity 1 Kit
Sample Volume Cell Culture Supernates (100uL),Tissue Lysates (10uL),Cerebrospinal Fluid (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.