Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Androgen R/NR3C4 Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Catalog No. IC58762G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC58762G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC58762G100 Supplier R&D Systems Supplier No. IC58762G100UG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Androgen R/NR3C4
Applications Flow Cytometry
Classification Monoclonal
Clone 2621E
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/106 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias AIS, androgen receptor, AndrogenR, AR, DHTR, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, SMAX1SBMA, spinal and bulbar muscular atrophy
Host Species Rabbit
Immunogen Synthetic Human Androgen R/NR3C4 peptide
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.