Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Androgen R/NR3C4 Isoform 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $409.00
Specifications
Antigen | Androgen R/NR3C4 |
---|---|
Clone | 1022602 |
Dilution | Immunocytochemistry 8-25 ug/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB58764SP
![]() |
R&D Systems
MAB58764SP |
25 μg |
Each for $200.50
|
|
|||||
MAB58764100
![]() |
R&D Systems
MAB58764100 |
100 μg |
Each for $409.00
|
|
|||||
Description
Androgen R/NR3C4 Monoclonal antibody specifically detects Androgen R/NR3C4 in Human samples. It is validated for ImmunocytochemistrySpecifications
Androgen R/NR3C4 | |
Immunocytochemistry 8-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
AIS, androgen receptor, AndrogenR, AR, DHTR, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, NR3C4, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SBMA, SMAX1, SMAX1SBMA, spinal and bulbar muscular atrophy | |
Human Androgen R/NR3C4 isoform 2 synthetic peptide, Accession # P10275 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1022602 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human | |
P10275 | |
367 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title