Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Angiopoietin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$200.50 - $2020.00
Specifications
Antigen | Angiopoietin-1 |
---|---|
Clone | 171733 |
Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9231SP
![]() |
R&D Systems
MAB9231SP |
25 μg |
Each for $200.50
|
|
|||||
MAB9231100
![]() |
R&D Systems
MAB9231100 |
100 μg |
Each for $549.50
|
|
|||||
MAB9231500
![]() |
R&D Systems
MAB9231500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Angiopoietin-1 Monoclonal specifically detects Angiopoietin-1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).Specifications
Angiopoietin-1 | |
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
Q15389 | |
284 | |
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-1 Ser20-Phe498 Accession # Q15389 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Angiopoietin-1 in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) Ang-2, rhAng-4, rmANGPTL3, or rhANGPTL7. |
171733 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, ANGPT1, KIAA0003 | |
ANGPT1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title