Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Angiopoietin-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB0984100
This item is not returnable.
View return policy
Description
Angiopoietin-2 Monoclonal specifically detects Angiopoietin-2 in Human samples. It is validated for ELISA.Specifications
Angiopoietin-2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
AGPT2, ANG2, ANG-2, angiopoietin 2, angiopoietin-2, angiopoietin-2a, angiopoietin-2B, angiopoitin 2, ANGPT2, Tie2-ligand | |
Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-2 Asp68-Phe496 Accession # O15123 | |
100 μg | |
285 | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
ELISA | |
85834R | |
ELISA | |
O15123 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Angiopoietin-2 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction