Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Angiopoietin-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9641SP
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9641SP 25 μg
MAB9641 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9641SP Supplier R&D Systems Supplier No. MAB9641SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Angiopoietin-4 Monoclonal specifically detects Angiopoietin-4 in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).

Specifications

Antigen Angiopoietin-4
Applications ELISA
Classification Monoclonal
Clone 156210
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y264
Gene Alias AGP4, ANG3, ANG-3, ANG-4, ANG4angiopoietin-3, angiopoietin 4, Angiopoietin-3, angiopoietin-4, ANGPT4, dJ824F16.2 (angiopoietin 4), MGC138181, MGC138183
Gene Symbols ANGPT4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Angiopoietin-4 Met1-Ile503 Accession # Q9Y264
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51378
Test Specificity Detects human Angiopoietin-4 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human (rh) Angiopoietin-1, -2, rhAngiopoietin-like 3, rhAngiopoietin-like 4, rhTie-2, or recombinant mouse Angiopoietin-3 was observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.