Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Angiopoietin-like 6, Carrier-Free Recombinant Protein

Catalog No. 8466AN025CF
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-66A-N025CF 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-66A-N025CF Supplier R&D Systems™ Supplier No. 8466AN025/CF

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human Angiopoietin-like Protein 6/ANGPTL6 (Catalog # 8466-AN/CF) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera (Catalog # 2078-T4 ) binds with an ED50 of 30.0-360 ng/mL.

Specifications

Accession Number Q8NI99
For Use With (Application) Bioactivity
Gene ID (Entrez) 83854
Molecular Weight (g/mol) Observed M.W.: 58-69kDa and 31-41kDa, reducing conditions, Theoretical M.W.: 51kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 6/ANGPTL6 protein Ala20-Leu470,with an N-terminal c-Myc tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias AGF, AGFAngiopoietin-related protein 5, Angiopoietin like Protein 6, angiopoietin-like 6, Angiopoietin-like protein 6, Angiopoietin-related growth factor, ANGPTL6, ARP5, ARP5angiopoietin-related protein 6
Common Name Angiopoietin-like Protein 6/ANGPTL6
Gene Symbol ANGPTL6
Biological Activity Measured by its ability to promote the expansion of E16 rat liver mononuclear cells in vitro, in the presence of Recombinant Mouse SCF/c-kit Ligand, Recombinant Mouse Thrombopoietin/Tpo, and Recombinant Mouse Flt-3 Ligand. The ED50 for this effect is typically 8-40ng/mL in the presence of a cross-linking Mouse Anti-Human c-Myc Monoclonal Antibody.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ala20-Leu470, with an N-terminal c-Myc tag
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.