Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Angiopoietin-like Protein 3/ANGPTL3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems MAB38292SP
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Monoclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot.Specifications
Angiopoietin-like Protein 3/ANGPTL3 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Angiopoietin-like Protein 3/ANGPTL3 Ile19-Glu460 Accession # Q9Y5C1 | |
25 μg | |
27329 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
388610 | |
Western Blot 1 ug/mL | |
Q9Y5C1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human Angiopoietin-like Protein 3/ANGPTL3 in direct ELISAs. | |
Human | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction