Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Angiopoietin-like Protein 3/ANGPTL3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3829
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3829 100 μg
AF3829SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3829 Supplier R&D Systems Supplier No. AF3829
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Angiopoietin-like Protein 3/ANGPTL3 Polyclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Angiopoietin-like Protein 3/ANGPTL3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9Y5C1
Gene Alias AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2
Gene Symbols ANGPTL3
Host Species Sheep
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Angiopoietin-like 3 (R&D Systems, Catalog # 3829-AN) Ser17-Glu460 Accession # Q9Y5C1
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27329
Test Specificity Detects human Angiopoietin-like 3 in direct ELISAs and Western blots. In direct ELISAs, less than 20% cross-reactivity with recombinant mouse Angiopoietin-like 3 is observed and less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-like 1 and rhAngiopoietin-like 4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.