Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Angiopoietin-like Protein 3/ANGPTL3 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF3829
This item is not returnable.
View return policy
Description
Angiopoietin-like Protein 3/ANGPTL3 Polyclonal specifically detects Angiopoietin-like Protein 3/ANGPTL3 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
Angiopoietin-like Protein 3/ANGPTL3 | |
Polyclonal | |
Biotin | |
Q9Y5C1 | |
ANGPTL3 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Angiopoietin-like 3 (R&D Systems, Catalog # 3829-AN) Ser17-Glu460 Accession # Q9Y5C1 | |
50 μg | |
Primary | |
Detects human Angiopoietin-like 3 in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-1, rhAngiopoietin-2, rhAngiopoietin-4, rhANGPTL4, recombinant mouse (rm) Angiopoietin-3, rmANGPTL3 and rmANGPTL4 is observed. | |
Human | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
AGNPT5, Ang-5, angiopoietin 5, Angiopoietin-5, angiopoietin-like 3, Angiopoietin-like protein 3, angiopoietin-related protein 3, ANGPT5angiopoietin-5, ANGPTL3, FHBL2 | |
Sheep | |
Affinity Purified | |
RUO | |
27329 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction