Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Angiopoietin-like Protein 7/ANGPTL7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $686.23
Specifications
| Antigen | Angiopoietin-like Protein 7/ANGPTL7 |
|---|---|
| Clone | 121408 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Angiopoietin-like Protein 7/ANGPTL7 Monoclonal specifically detects Angiopoietin-like Protein 7/ANGPTL7 in Human samples. It is validated for Western Blot.Specifications
| Angiopoietin-like Protein 7/ANGPTL7 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| angiopoietin-like 7, Angiopoietin-like factor, Angiopoietin-like protein 7, angiopoietin-related protein 7, ANGPTL7, AngX, CDT6, CDT6angiopoietin-like factor (CDT6), Cornea-derived transcript 6 protein, dJ647M16.1, RP4-647M16.2 | |
| ANGPTL7 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 121408 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O43827 | |
| 10218 | |
| E. coli-derived recombinant human Angiopoietin-like Protein 7/ANGPTL7 Lys28-Pro346 Accession # O43827 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Angiopoietin-like Protein 7/ANGPTL7 in direct ELISAs and Western blots. In Western blots, less than 3% cross-reactivity with recombinant human Angiopoietin-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title