Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Angiostatin Kringle 1-3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB926
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB926 500 μg
MAB926SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB926 Supplier R&D Systems Supplier No. MAB926
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Angiostatin Monoclonal specifically detects Angiostatin in Human samples. It is validated for Western Blot.

Specifications

Antigen Angiostatin
Applications Western Blot
Classification Monoclonal
Clone 79735
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P00747
Gene Symbols PLG
Host Species Mouse
Immunogen E. coli-derived recombinant human Angiostatin Kringle 1-3 Val98-Ser358 (Cys88Ser, Cys316Ser) Accession # P00747
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5340
Test Specificity Detects human Angiostatin Kringle 1-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody also detects plasminogen but does not cross-react with recombinant mouse Angiostatin.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.