Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Annexin A10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3544
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3544 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF3544 Supplier R&D Systems Supplier No. AF3544
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Annexin A10 Polyclonal specifically detects Annexin A10 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Annexin A10
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q9UJ72
Gene Alias annexin 14, annexin A10, annexin-10, annexin-14, ANX14, ANX14Annexin-10, ANXA10
Gene Symbols ANXA10
Host Species Goat
Immunogen E. coli-derived recombinant human Annexin A10 Met1-Tyr324 Accession # Q9UJ72
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11199
Test Specificity Detects endogenous human Annexin A10 in Western blots. In Western blots, this antibody does not cross-react with recombinant human Annexin A1, A2, A3, A4, A6, A7, A8, A9, A11, or A13.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.