Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Annexin A4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4146SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4146SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4146SP Supplier R&D Systems Supplier No. MAB4146SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Annexin A4 Monoclonal specifically detects Annexin A4 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Annexin A4
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 694624
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P09525
Gene Alias AIV, annexin A4, Annexin IV, annexin IV (placental anticoagulant protein II), annexin-4, ANX4, ANX4Carbohydrate-binding protein p33/p41, ANXA4, Chromobindin-4, chromobindin-4,35-beta calcimedin, DKFZp686H02120, Endonexin I, Lipocortin IV, MGC75105, P32.5, PAP-II, PIG28, Placental anticoagulant protein II, PP4-X, proliferation-inducing gene 28, proliferation-inducing protein 28, Protein II, ZAP36
Gene Symbols ANXA4
Host Species Mouse
Immunogen E. coli-derived recombinant human Annexin A4 Met1-Asp319 Accession # P09525
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307
Test Specificity Detects human Annexin A4 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Annexin A1, A2, A3, A5, A6, A8, A9, A10, A11, or 13A is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.