Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Annexin A6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5186
This item is not returnable.
View return policy
Description
Annexin A6 Monoclonal specifically detects Annexin A6 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Annexin A6 | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
P08133 | |
ANXA6 | |
E. coli-derived recombinant human Annexin A6 Ala2-Asp673 Accession # P08133 | |
100 μg | |
Primary | |
Detects human Annexin A6 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human Annexin A1, A2, A3, A4, A8, A9, A10, A11, or A13 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
686921 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
67 kDa calelectrin, annexin A6, Annexin VI, annexin VI (p68), annexin-6, ANX6, ANXA6, calcium-binding protein p68, calelectrin, calphobindin II, calphobindin-II, CBP68, Chromobindin-20, CPB-II, Lipocortin VI, p68, p70, Protein III | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
309 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction