Learn More
Human Annexin A8/ANXA8 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Annexin A8/ANXA8 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P13928 |
| Gene Alias | annexin A8, Annexin VIII, Annexin-8, Anx8, ANXA8, ANXA8L2, FLJ53095, VAC-beta, Vascular anticoagulant-beta |
| Gene Symbols | ANXA8 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.