Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridium botulinum Toxin F, Human, Monoclonal Antibody, Abnova™

Catalog No. 89135968 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-968 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-968 Supplier Abnova Corporation Supplier No. MAB5419

human monoclonal antibody raised against full-length recombinant Clostridium Botulinum Toxin F.

Specifications

Antigen Clostridium Botulinum Toxin F
Applications Dot Blot, ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Recombinant antibody raised against full length recombinant Clostridium Botulinum Toxin F.
Dilution ELISA (1:1000-1:2000) Western Blot (1-5 ug/mL) Dot Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Human
Immunogen Recombinant myc and His fusion protein corresponding to Clostridium Botulinum Toxin F.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody specifically detects Toxin F as determined from testing bacterial samples in Dot Blot and ELISA assays.
Target Species Bacteria
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.