Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Escherichia coli O157:H7, Human, Monoclonal Antibody, Abnova™
human monoclonal antibody raised against full-length recombinant Escherichia coli O157:H7.
Supplier: Abnova Corporation MAB5413
Specifications
Escherichia coli O157:H7 | |
Monoclonal | |
Recombinant antibody raised against full length recombinant Escherichia coli O157:H7. | |
In PBS | |
Recombinant myc and His fusion protein corresponding to Escherichia coli O157:H7. | |
RUO | |
It specifically detects protein derived from testing bacterial sample at various concentrations. See image Below. | |
Store at 4°C. For long term storage store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Dot Blot, ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000-1:2000) Western Blot (1-5 ug/mL) Dot Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user. | |
E. coli | |
100 μg | |
Primary | |
Bacteria | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction