Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human AP-2 beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5634
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5634 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5634 Supplier R&D Systems Supplier No. AF5634
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

AP-2 beta/TFAP2B Polyclonal specifically detects AP-2 beta/TFAP2B in Human samples. It is validated for Western Blot.

Specifications

Antigen AP-2 beta/TFAP2B
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q92481
Gene Alias activating enhancer binding protein 2 beta, Activating enhancer-binding protein 2-beta, AP2 beta, AP-2B, AP2-B, AP2-beta, CHAR, MGC21381, TFAP2B, transcription factor AP-2 beta (activating enhancer binding protein 2 beta), transcription factor AP-2 beta (activating enhancer-binding protein 2 beta), transcription factor AP-2-beta
Gene Symbols TFAP2B
Host Species Goat
Immunogen E. coli-derived recombinant human AP-2 beta Leu121-Phe218 Accession # Q92481
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7021
Test Specificity Detects endogenous human AP-2 beta in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.