Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APBA3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5689
This item is not returnable.
View return policy
Description
APBA3/Mint-3 Polyclonal specifically detects APBA3/Mint-3 in Human samples. It is validated for Western Blot.Specifications
APBA3/Mint-3 | |
Polyclonal | |
Unconjugated | |
O96018 | |
APBA3 | |
E. coli-derived recombinant human APBA3 Met1-Leu138 Accession # O96018 | |
100 μg | |
Primary | |
Detects human APBA3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) X11-alpha and rhX11-beta is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Adapter protein X11gamma, amyloid beta (A4) precursor protein-binding, family A, member 3, amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2), MGC:15815, mint3, Mint-3, Neuronal Munc18-1-interacting protein 3, Neuron-specific X11L2 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, X11L2, X11L2amyloid beta A4 precursor protein-binding family A member 3, X11-like 2 protein | |
Sheep | |
Affinity Purified | |
RUO | |
9546 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction