Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APCDD1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB10501R100UG
This item is not returnable.
View return policy
Description
APCDD1 Monoclonal specifically detects APCDD1 in Human samples. It is validated for Flow Cytometry.Specifications
APCDD1 | |
Monoclonal | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, APC Down Regulated 1, B7323, DRAPC1, HYPT1, protein APCDD1 | |
Mouse myeloma cell line, NS0-derived human APCDD1, Leu27-His492, Accession # Q8J025 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
2584B | |
Flow Cytometry 0.25-1 ug/106 cells | |
Q8J025 | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
147495 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction