Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APH1A Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6908
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6908 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6908 Supplier R&D Systems Supplier No. MAB6908
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APH1A Monoclonal specifically detects APH1A in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen APH1A
Applications Immunohistochemistry
Classification Monoclonal
Clone 665404
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96BI3
Gene Alias 6530402N02Rik, anterior pharynx defective 1 homolog A (C. elegans), APH-1, APH-1A, aph-1alpha, CGI-78, gamma-secretase subunit APH-1A, Presenilin-stabilization factor, PSF
Gene Symbols APH1A
Host Species Mouse
Immunogen Human APH1A synthetic peptide SSFERFEIFPKHVTDRSDARLQY Accession # Q96BI3
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51107
Test Specificity Detects human APH1A in direct ELISAs.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.