Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APLP-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Catalog No. FAB3129G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3129G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3129G100 Supplier R&D Systems Supplier No. FAB3129G100UG
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APLP-1 Monoclonal specifically detects APLP-1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen APLP-1
Applications Flow Cytometry
Classification Monoclonal
Clone 354008
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P51693
Gene Alias amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1
Gene Symbols APLP1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human APLP-1 Gln34-Glu580 (predicted) Accession # P51693
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333
Test Specificity Detects human APLP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse APLP-1 is observed and no cross-reactivity with recombinant human APLP-2 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.