Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APLP-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB49451
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB49451 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB49451 Supplier R&D Systems Supplier No. MAB49451
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APLP-2 Monoclonal specifically detects APLP-2 in Human samples. It is validated for Western Blot.

Specifications

Antigen APLP-2
Applications Western Blot
Classification Monoclonal
Clone 397702
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q06481
Gene Alias amyloid beta (A4) precursor-like protein 2, amyloid precursor protein homolog HSD-2, Amyloid protein homolog, amyloid-like protein 2, APLP2, APLP-2, APPH, APPHCDEI box-binding protein, APPL2, CDEBP, WALPLP2, YWK-II
Gene Symbols APLP2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human APLP-2 Leu30-Ser680 Accession # Q06481
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 334
Test Specificity Detects human APLP-2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse APLP-2 or recombinant human APLP-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.