Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein A-I/ApoA1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems MAB36641100
This item is not returnable.
View return policy
Description
Apolipoprotein A-I/ApoA1 Monoclonal specifically detects Apolipoprotein A-I/ApoA1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.Specifications
| Apolipoprotein A-I/ApoA1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| Alp-1, APOA1, Apo-AI, ApoA-I, Apolipoprotein A1, apolipoprotein A-I, Apolipoprotein AI, Brp-14, Ltw-1, Lvtw-1, MGC117399, Sep-1, Sep-2 | |
| E. coli-derived recombinant human ApoA1 Asp25-Gln267 Accession # P02647 | |
| 100 μg | |
| Primary | |
| Detects human ApoA1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 2083A | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-25 ug/mL | |
| P02647 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 335 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction