Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Apolipoprotein A-II/ApoA2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4215
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4215 100 μg
MAB4215SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4215 Supplier R&D Systems Supplier No. MAB4215
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Apolipoprotein A-II/ApoA2 Monoclonal specifically detects Apolipoprotein A-II/ApoA2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Apolipoprotein A-II/ApoA2
Applications Western Blot
Classification Monoclonal
Clone 395906
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P02652
Gene Alias Alp-2, APOA2, apoAII, Apo-AII, ApoA-II, Apolipoprotein A2, apolipoprotein A-II, Apolipoprotein AII, Hdl-1
Gene Symbols APOA2
Host Species Rat
Immunogen E. coli-derived recombinant human ApoA2 Gln24-Gln100 Accession # P02652
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 336
Test Specificity Detects human ApoA2 in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) ApoA1 or rhApoE.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.