Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein A-IV/ApoA4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $491.00
Specifications
Antigen | Apolipoprotein A-IV/ApoA4 |
---|---|
Clone | 988829 |
Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB8125SP
![]() |
R&D Systems
MAB8125SP |
25 μg |
Each for $200.50
|
|
|||||
MAB8125100
![]() |
R&D Systems
MAB8125100 |
100 μg |
Each for $491.00
|
|
|||||
Description
Apolipoprotein A-IV/ApoA4 Monoclonal specifically detects Apolipoprotein A-IV/ApoA4 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Apolipoprotein A-IV/ApoA4 | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Monoclonal | |
Supernatant | |
Human | |
P06727 | |
337.0 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
988829 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
apo-AIV, ApoA-IV, Apolipoprotein A4, apolipoprotein A-IV, Apolipoprotein AIV, MGC142154, MGC142156 | |
E. coli-derived human Apolipoprotein A-IV/ApoA4 protein, Glu21-Ser396, Accession No. P06727 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Apolipoprotein A-IV/ApoA4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title