Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein B100/ApoB100 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3260SP
This item is not returnable.
View return policy
Description
Apolipoprotein B100/ApoB100 Polyclonal specifically detects Apolipoprotein B100/ApoB100 in Human samples. It is validated for Western Blot.Specifications
Apolipoprotein B100/ApoB100 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APOB | |
E. coli-derived recombinant human ApoB100 Ile3374-Val3600 Accession # AAP72970 | |
25 μg | |
Primary | |
Detects human ApoB100 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
AAP72970 | |
Goat | |
Affinity Purified | |
RUO | |
338 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction