Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Apolipoprotein C-II/ApoC2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | Apolipoprotein C-II/ApoC2 |
---|---|
Clone | 2554B |
Dilution | ELISA, Immunohistochemistry 0.3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
Applications | ELISA, Immunohistochemistry, Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB44971SP
![]() |
R&D Systems
MAB44971SP |
25 μg |
Each for $200.50
|
|
|||||
MAB44971100
![]() |
R&D Systems
MAB44971100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Apolipoprotein C-II/ApoC2 Monoclonal specifically detects Apolipoprotein C-II/ApoC2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry.Specifications
Apolipoprotein C-II/ApoC2 | |
ELISA, Immunohistochemistry 0.3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
P02655 | |
344 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
2554B | |
ELISA, Immunohistochemistry, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
APC2, APOC2, Apo-CII, ApoC-II, Apolipoprotein C2, apolipoprotein C-II, Apolipoprotein CII, MGC75082 | |
E. coli-derived human Apolipoprotein C-II/ApoC2, Thr23-Glu101, Accession # P02655 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title