Learn More
Human Apolipoprotein C-II/ApoC2 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Apolipoprotein C-II/ApoC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL |
| Gene Accession No. | P02655 |
| Gene Alias | APC2, APOC2, Apo-CII, ApoC-II, Apolipoprotein C2, apolipoprotein C-II, Apolipoprotein CII, MGC75082 |
| Gene Symbols | APOC2 |
| Host Species | Sheep |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.