Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APP/Protease Nexin II Pan Specific Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1168SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1168SP 25 μg
AF1168 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF1168SP Supplier R&D Systems Supplier No. AF1168SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

APP Polyclonal specifically detects APP in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen APP
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_958817
Gene Alias Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II
Gene Symbols APP
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP 695 +1 frameshift mutant Leu18-Arg336 Accession # NP_958817
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351
Test Specificity Detects human APP695 +1 and human APP770 in direct ELISAs and Western blots. This antibody recognizes a common epitope present on APP695 +1 and APP770 found within the segments Leu18 - Arg288 or Pro365 - Arg411 of APP770 (Accession P05067).
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.