Learn More
Human APP/Protease Nexin II (Research Grade Aducanumab Biosimilar) Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Supplier:  R&D Systems FAB9710U100UG
Catalog No. FAB9710U100
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
APP Monoclonal specifically detects APP in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
APP
Monoclonal
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
Mouse
100 μg
351
Human
IgG1
Flow Cytometry
Hu15
Flow Cytometry 0.25-1 μg/106 cells
Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II
Human APP/Protease Nexin II
Primary
Detects human APP/Protease Nexin II based on Aducanumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Aducanumab. This product is for research use only.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.