Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APP/Protease Nexin II (Research Grade Aducanumab Biosimilar) Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9710R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9710R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9710R100 Supplier R&D Systems Supplier No. FAB9710R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APP Monoclonal specifically detects APP in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen APP
Applications Flow Cytometry
Classification Monoclonal
Clone Hu15
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide.
Gene Alias Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II
Host Species Mouse
Immunogen Human APP/Protease Nexin II
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 351.0
Test Specificity Detects human APP/Protease Nexin II based on Aducanumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Aducanumab. This product is for research use only.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.