Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APP/Protease Nexin II (Research Grade Aducanumab Biosimilar) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9710SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9710SP 25 μg
MAB9710100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9710SP Supplier R&D Systems Supplier No. MAB9710SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Monoclonal Antibody

APP Monoclonal specifically detects APP in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen APP
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone Hu15
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II
Host Species Human
Immunogen Human APP/Protease Nexin II
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 351
Test Specificity Detects human APP/Protease Nexin II based on Aducanumab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Aducanumab. This product is for research use only.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.