Learn More
Human APP/Protease Nexin II (Research Grade Solanezumab Biosimilar) Alexa Fluor 750-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | APP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | Hu13 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Flow Cytometry 0.25-1μg/106 cells |
| Formulation | Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide. |
| Gene Alias | Amyloid beta, amyloid beta (A4) precursor protein-binding, family B, member 2, amyloid beta A4 precursor protein-binding family B member 2, Amyloid beta precursor protein, APP, beta Amyloid, Protease Nexin II |
| Host Species | Human |
| Immunogen | Human APP/Protease Nexin II |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.