Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APP695+1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $585.50
Specifications
Antigen | APP 695+1 |
---|---|
Clone | 128715 |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
APP 695+1 Monoclonal specifically detects APP 695+1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
APP 695+1 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
351 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP695 +1 frameshift mutant Leu18-Arg336 (predicted) Accession # NP_958817 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human APP695 +1 in Western blots. It recognizes a common epitope present on APP695 +1 and APP770 found within the segments L18-R288 or P365-R411 of APP770 (Accession # P05067). |
128715 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
NP_958817 | |
APP | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title