Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human APP695+1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF890SP
This item is not returnable.
View return policy
Description
APP 695+1 Polyclonal specifically detects APP 695+1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
APP 695+1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APP | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human APP695 +1 frameshift mutant Met1-Arg336 Accession # NP_958817 | |
25 μg | |
Primary | |
Detects human APP695 +1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
NP_958817 | |
Goat | |
Affinity Purified | |
RUO | |
351 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction