Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human APRIL/TNFSF13 Alexa Fluor 750-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC8843S100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC8843S100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC8843S100 Supplier R&D Systems Supplier No. IC8843S100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

APRIL/TNFSF13 Monoclonal specifically detects APRIL/TNFSF13 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen APRIL/TNFSF13
Applications Flow Cytometry
Classification Monoclonal
Clone 670840
Conjugate Alexa Fluor 750
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O75888
Gene Alias A proliferation-inducing ligand, APRILFLJ57090, CD256, CD256 antigen, TALL2, TALL-2, TNF- and APOL-related leukocyte expressed ligand 2, TNF-related death ligand 1, TNFSF13, TRDL1, TRDL-1, tumor necrosis factor (ligand) superfamily, member 13, tumor necrosis factor ligand superfamily member 13, tumor necrosis factor-like protein ZTNF2, tumor necrosis factor-related death ligand-1, ZTNF2
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293EBNA-derived recombinant human APRIL/TNFSF13 Ala105-Leu250 Accession # O75888
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8741
Test Specificity Detects human APRIL/TNFSF13 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse APRIL, recombinant human (rh) BAFF, rhEDA-A2, rhEDA, rhFas Ligand, rhLIGHT, rhLT a1/b2, rhLT a2/b1, rhOX40 Ligand, rhTNF alpha, rhTRAIL, rhTRANCE, rh4-1BB Ligand, rhVEGI, or rhGITR Ligand is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.